Learn More
Medchemexpress LLC HY-12364 10mg Medchemexpress, C75 CAS:218137-86-1 Purity:>98%
SDP
Catalog No. 502028877
Encompass_Preferred
Medchemexpress, HY-12364 10mg C75 CAS:218137-86-1 C75 is a synthetic fatty-acid synthase (FASN) inhibitor; inhibits prostate cancer cells PC3 with an IC50 of 35 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8877 Supplier Medchemexpress LLC Supplier No. HY1236410MG
Medchemexpress, HY-12364 10mg C75 CAS:218137-86-1 C75 is a synthetic fatty-acid synthase (FASN) inhibitor; inhibits prostate cancer cells PC3 with an IC50 of 35 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.