Learn More
Medchemexpress LLC HY-12364 5mg Medchemexpress, C75 CAS:218137-86-1 Purity:>98%
SDP
Catalog No. 501932582
Encompass_Preferred
Medchemexpress, HY-12364 5mg C75 CAS:218137-86-1 C75 is a synthetic fatty-acid synthase ( FASN ) inhibitor; inhibits prostate cancer cells PC3 with an IC 50 of 35 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2582 Supplier Medchemexpress LLC Supplier No. HY123645MG
Medchemexpress, HY-12364 5mg C75 CAS:218137-86-1 C75 is a synthetic fatty-acid synthase ( FASN ) inhibitor; inhibits prostate cancer cells PC3 with an IC 50 of 35 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.