Learn More
Selleck Chemical LLC C75-5mg
SDP
Catalog No. 502389956
Encompass_Preferred
C75 (4-Methylene-2-octyl-5-oxotetrahydrofuran-3-carboxylic acid) , a more stable synthetic mimic of cerulenin, inhibits fatty-acid synthase (FAS/FASN) with an IC50 of 15.53 μM and has potential therapeutic effects in several cancer models. C75 is also a potent CPT1A activator.
Catalog No. 50-238-9956 Supplier Selleck Chemical LLC Supplier No. S98195MG
C75 (4-Methylene-2-octyl-5-oxotetrahydrofuran-3-carboxylic acid) , a more stable synthetic mimic of cerulenin, inhibits fatty-acid synthase (FAS/FASN) with an IC50 of 15.53 μM and has potential therapeutic effects in several cancer models. C75 is also a potent CPT1A activator.