Learn More
Cayman Chemical C75 Fatty Acids and Derivative
SDP
Supplier:  Cayman Chemical 1000527010 MG
Encompass_Preferred
An anorexigenic inhibitor of FASN; leads to profound weight loss and feeding inhibition in both high-fat diet wild type obese and leptin-deficient ob/ob mice; cytotoxic to many human cancer cell lines
Catalog No. 50-289-0753