Learn More
Selleck Chemical LLC C75 trans S8915-10MM/1ML
SDP
Catalog No. 5000024218
Encompass_Preferred
C75 trans (trans-C75 ( )-C75) is a novel potent synthetic inhibitor of fatty acid synthase (FAS/FASN) with IC50 of 35 M and 50 M in clonogenic assays and spheroid growth assay respectively C75 is used as a tool for studying fatty acid synthesis in metabolic disorders and cancer
Catalog No. 50-000-24218 Supplier Selleck Chemical LLC Supplier No. S891510MM/1ML
C75 trans (trans-C75 ( )-C75) is a novel potent synthetic inhibitor of fatty acid synthase (FAS/FASN) with IC50 of 35 M and 50 M in clonogenic assays and spheroid growth assay respectively C75 is used as a tool for studying fatty acid synthesis in metabolic disorders and cancer