Learn More
Sigma Aldrich Fine Chemicals Biosciences C7BzO1G
Catalog No. 502901215
Encompass
3-(4-heptyl)phenyl 3-hydroxypropyl dimethyl ammonio propane sulfonate (C7BzO) is a zwitterionic detergent comprising of a zwitterionic hydrophilic head connected to the hydrophobic tail through a linker region. The prelinker region enhances detergent solubility. C7BzO is effective in sample preparation from plant membranes.
Catalog No. 50-290-1215 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C08561G
3-(4-heptyl)phenyl 3-hydroxypropyl dimethyl ammonio propane sulfonate (C7BzO) is a zwitterionic detergent comprising of a zwitterionic hydrophilic head connected to the hydrophobic tail through a linker region. The prelinker region enhances detergent solubility. C7BzO is effective in sample preparation from plant membranes.