Learn More
Aviva Systems Biology Corp C7orf16 Antibody - middle region (ARP52229_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52229_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C7orf16. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C7orf16. Alternate names: GSBS, C7orf16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-665