Learn More
Aviva Systems Biology Corp C7orf38 Antibody - middle region (ARP55465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C7orf38. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C7orf38. Alternate names: DKFZp727G131, FLJ36794, C7orf38ApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-61514