Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C8-C40 Alkanes Calibration Standard in Dichloromethane (varied), MilliporeSigma™ Supelco™

Catalog No. 111017381 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7381 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7381 Supplier MilliporeSigma Supelco Supplier No. 40147U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Qualitative standard. May require warming before use.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) Octane 500 μg/mL; Nonane 500 μg/mL; Decane 500 μg/mL; Undecane 500 μg/mL; Dodecane 500 μg/mL; Tridecane 500 μg/mL; Tetradecane 500 μg/mL; Pentadecane 500 μg/mL; Hexadecane 500 μg/mL; Heptadecane 500 μg/mL; Octadecane 500 μg/mL; Nonadecane 500 μg/mL; Pristane 500 μg/mL; Eicosane 500 μg/mL; Phytane 500 μg/mL; Docosane 500 μg/mL; Tricosane 500 μg/mL; Tetracosane 500 μg/mL; Pentacosane 500 μg/mL; Heneicosane 500 μg/mL; Hexacosane 500 μg/mL; Heptacosane 500 μg/mL; Octacosane 500 μg/mL; Nonacosane 500 μg/mL; Triacontane 500 μg/mL; Hentriacontane 500 μg/mL; Dotriacontane 500 μg/mL; Tritriacontane 500 μg/mL; Tetratriacontane 500 μg/mL; Pentatriacontane 500 μg/mL; Hexatriacontane 500 μg/mL; Heptatriacontane 500 μg/mL; Octatriacontane 500 μg/mL; Nonatriacontane; Tetracontane; Carbon disulfide 5000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.