Learn More
Cayman Chemical C8 Ceramide-1-phosphate (d18:1/8:0) (ammonium salt), Purity: ≥90%, CAS: 474943-70-9, 25 mg
SDP
Catalog No. 502894414
Encompass_Preferred
C8 ceramide-1-phosphate is a short-chain analog of the naturally occurring ceramide-1-phosphate. The effects of C8 ceramide-1-phosphate are essentially the opposite of C8 ceramide; it induces DNA synthesis and cell division. The effects of C8 ceramide-1-phosphate can be attenuated by simultaneous treatment with C2 or C8 ceramide.{5183}
Catalog No. 50-289-4414 Supplier Cayman Chemical Supplier No. 6254725 MG
C8 ceramide-1-phosphate is a short-chain analog of the naturally occurring ceramide-1-phosphate. The effects of C8 ceramide-1-phosphate are essentially the opposite of C8 ceramide; it induces DNA synthesis and cell division. The effects of C8 ceramide-1-phosphate can be attenuated by simultaneous treatment with C2 or C8 ceramide.{5183}