Learn More
Enzo Life Sciences C8 Ceramide (25 mg). CAS: 74713-59-0
SDP
Supplier:  Enzo Life Sciences BMLSL1120025
Encompass_Preferred
A biologically active, cell-permeable, but nonphysiologic ceramide analog. It induces phosphorylation of epidermal growth factor receptor on Thr-669 in A-431 cells by stimulation of ceramide-activated protein kinase and induces apoptosis. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2108