Learn More
Medchemexpress LLC C8-Ceramide (N-Octanoyl-D-erythro-sphingosine) | 74713-59-0 | 98.0% | 425.69 | 1 G
SDP
Catalog No. 5000346869
Encompass_Preferred
C8-Ceramide (N-Octanoyl-D-erythro-sphingosine) is a cell-permeable analog of naturally occurring ceramides. It exhibits anti-proliferation properties and functions as a potent chemotherapeutic agent. It stimulates dendritic cells to promote T cell responses during virus infections and induces slight activation of protein kinase (PKC) in vitro.
  • Cell-permeable analog of ceramides
  • Anti-proliferation and chemotherapeutic agent
  • Stimulates dendritic cells for T cell responses
  • Activates protein kinase C
  • Reduces tumor-cell proliferation
  • Induces necrosis-like cell death
  • Increases ROS levels
  • Triggers apoptosis and G1 arrest in NSCLC H1299 cells
  • Enhances CD8+ and CD4+ T cell responses in viral infections
Catalog No. 50-003-46869 Supplier Medchemexpress LLC Supplier No. HY10839150MG
C8-Ceramide (N-Octanoyl-D-erythro-sphingosine) is a cell-permeable analog of naturally occurring ceramides. It exhibits anti-proliferation properties and functions as a potent chemotherapeutic agent. It stimulates dendritic cells to promote T cell responses during virus infections and induces slight activation of protein kinase (PKC) in vitro.
  • Cell-permeable analog of ceramides
  • Anti-proliferation and chemotherapeutic agent
  • Stimulates dendritic cells for T cell responses
  • Activates protein kinase C
  • Reduces tumor-cell proliferation
  • Induces necrosis-like cell death
  • Increases ROS levels
  • Triggers apoptosis and G1 arrest in NSCLC H1299 cells
  • Enhances CD8+ and CD4+ T cell responses in viral infections