Learn More
Cayman Chemical C8 CeramInd18 18 0 10mg
SDP
Catalog No. 502896480
Encompass_Preferred
An analog of C8 ceramide in which the carbonyl group of the ceramide is replaced by a methylene group; induces maximal DNA fragmentation in U937 cells after 6 hours of incubation (10 µM)
Catalog No. 50-289-6480 Supplier Cayman Chemical Supplier No. 6254510 MG
An analog of C8 ceramide in which the carbonyl group of the ceramide is replaced by a methylene group; induces maximal DNA fragmentation in U937 cells after 6 hours of incubation (10 µM)