Learn More
Enzo Life Sciences C8 ceramine, D-erythro (5 mg). CAS: 170926-06-4
SDP
Catalog No. 502012121
Encompass_Preferred
C8 ceramine is an analog of ceramide in which the amide is substituted with an amine. This substitution renders it inert to ceramidases. It is highly effective at induction of apoptosis in U937 cells. Ceramine induces peak DNA fragmentation in 6 hrs compared to 18 hrs for C8 ceramide. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2121 Supplier Enzo Life Sciences Supplier No. BMLSL1450005
C8 ceramine is an analog of ceramide in which the amide is substituted with an amine. This substitution renders it inert to ceramidases. It is highly effective at induction of apoptosis in U937 cells. Ceramine induces peak DNA fragmentation in 6 hrs compared to 18 hrs for C8 ceramide. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml). Long Term Storage: -20°C.