Learn More
Enzo Life Sciences C8-Dihydroceramide (25 mg). CAS: 145774-33-0
SDP
Catalog No. 502012110
Encompass_Preferred
C8 Dihydroceramide is inactive and may be used as a negative control for C8 ceramide. Purity: ≥98% (TLC). Solubility: Soluble in DMF (25 mg/ml), DMSO (5 mg/ml) or ethanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2110 Supplier Enzo Life Sciences Supplier No. BMLSL1130025
C8 Dihydroceramide is inactive and may be used as a negative control for C8 ceramide. Purity: ≥98% (TLC). Solubility: Soluble in DMF (25 mg/ml), DMSO (5 mg/ml) or ethanol (10mg/ml). Long Term Storage: -20°C.