Learn More
AdipoGen C8-DL-HSL
SDP
Catalog No. 502560301
Encompass_Preferred
Chemical. CAS 106983-30-6. Formula C12H21NO3. MW 227.3. Synthetic. N-Octanyol-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-0301 Supplier AdipoGen Supplier No. CDXO0062M010
Chemical. CAS 106983-30-6. Formula C12H21NO3. MW 227.3. Synthetic. N-Octanyol-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.