Learn More
Cayman Chemical C8 Galactosylceramide (d18:1/8:0), Purity: ≥98%, CAS: 41613-16-5, 10 mg
SDP
Catalog No. 503016026
Encompass_Preferred
A synthetic bioactive sphingolipid; increases the amount delivered and toxicity of doxorubicin in cancerous but not non-cancerous cells when incorporated in the nanoliposomal membrane of nanoliposomal-doxorubicin; induces proliferation and cytokine production by splenocytes in vitro (100-1,000 ng/ml); activates NF-κB production in C6 glioma cells (10 μM)
Catalog No. 50-301-6026 Supplier Cayman Chemical Supplier No. 2434610 MG
A synthetic bioactive sphingolipid; increases the amount delivered and toxicity of doxorubicin in cancerous but not non-cancerous cells when incorporated in the nanoliposomal membrane of nanoliposomal-doxorubicin; induces proliferation and cytokine production by splenocytes in vitro (100-1,000 ng/ml); activates NF-κB production in C6 glioma cells (10 μM)