Learn More
YMC America, Inc C8 guard 120A 20um 10x10mm
SDP
Catalog No. 5000880857
Encompass
YMC-Pack C8 columns are prepared using exhaustive bonding of a monomeric octylsilane to totally porous and spherical silica gels The functionalized silica is then treated with an endcapping process which produces a moderate 10% carbon loading on the standard 120 pore material Available in three pore sizes YMC-Pack C8 columns will separate many classes of compounds including pharmaceuticals organic chemicals and biologics
Catalog No. 50-008-80857 Supplier YMC America, Inc Supplier No. OC12S210110CC
YMC-Pack C8 columns are prepared using exhaustive bonding of a monomeric octylsilane to totally porous and spherical silica gels The functionalized silica is then treated with an endcapping process which produces a moderate 10% carbon loading on the standard 120 pore material Available in three pore sizes YMC-Pack C8 columns will separate many classes of compounds including pharmaceuticals organic chemicals and biologics