Learn More
YMC America, Inc C8-HG col 120A 10um 250x30mm
SDP
Catalog No. 5000879486
Encompass
YMC-Pack C8 columns are prepared using exhaustive bonding of a monomeric octylsilane to totally porous and spherical silica gels The functionalized silica is then treated with an endcapping process which produces a moderate 10% carbon loading on the standard 120 pore material Available in three pore sizes YMC-Pack C8 columns will separate many classes of compounds including pharmaceuticals organic chemicals and biologics
Catalog No. 50-008-79486 Supplier YMC America, Inc Supplier No. OCG12S112530WT
YMC-Pack C8 columns are prepared using exhaustive bonding of a monomeric octylsilane to totally porous and spherical silica gels The functionalized silica is then treated with an endcapping process which produces a moderate 10% carbon loading on the standard 120 pore material Available in three pore sizes YMC-Pack C8 columns will separate many classes of compounds including pharmaceuticals organic chemicals and biologics