Learn More
AdipoGen C8-L-HSL
SDP
Supplier:  AdipoGen CDXO0038M005
Encompass_Preferred
Chemical. CAS 147852-84-4. Formula C12H21NO3. MW 227.3. Synthetic. N-Octanoyl-L-homoserine lactone C8-HSL is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The applications of this molecule include infection prevention and regulation of virulence in general and in cystic fibrosis.
Catalog No. 50-256-0335