Learn More
Avanti Polar Lipids C8 PEG2000 CERAMIDE-25MG
Catalog No. NC2308313
Encompass
C8 PEG2000 Ceramide, 25MG, CAS# 212116-76-2, Purity: >99%
Catalog No. NC2308313 Supplier Avanti Polar Lipids Supplier No. 880170P25MG
C8 PEG2000 Ceramide, 25MG, CAS# 212116-76-2, Purity: >99%