Learn More
Medchemexpress LLC C8 PEG750 ceramide | 212116-76-2 | 99.9% | 750.0 g/mol (average) | (C2H4O)nC31H57NO6 | 5 MG
SDP
Catalog No. 5000222229
Encompass_Preferred
C8 PEG750 ceramide is a PEGylated ceramide lipid used to form lipid bilayer carriers such as liposomes and lipid nanoparticles for selective delivery of diagnostic and therapeutic agents to acidic diseased cells. It is provided as a high-purity research reagent suitable for formulation development and analytical studies.
  • High purity for reliable formulation and analysis.
  • PEGylated ceramide structure for steric stabilization of lipid assemblies.
  • Average molecular weight ~750 supports PEG shielding properties.
  • Suitable for liposome and LNP formulation development.
  • Available in small milligram pack sizes for research use.
Catalog No. 50-002-22229 Supplier Medchemexpress LLC Supplier No. HY144008B5MG
C8 PEG750 ceramide is a PEGylated ceramide lipid used to form lipid bilayer carriers such as liposomes and lipid nanoparticles for selective delivery of diagnostic and therapeutic agents to acidic diseased cells. It is provided as a high-purity research reagent suitable for formulation development and analytical studies.
  • High purity for reliable formulation and analysis.
  • PEGylated ceramide structure for steric stabilization of lipid assemblies.
  • Average molecular weight ~750 supports PEG shielding properties.
  • Suitable for liposome and LNP formulation development.
  • Available in small milligram pack sizes for research use.