Learn More
Medchemexpress LLC C8 PEG750 Ceramide | 212116-76-2 | 99.94% | 1 MG
SDP
Catalog No. 5000356204
Encompass_Preferred
C8 PEG750 Ceramide is a lipid product designed to synthesize a lipid bilayer carrier. It enables the selective delivery of various diagnostic and therapeutic agents to acidic diseased cells.
  • Synthesizes a lipid bilayer carrier
  • Facilitates selective delivery of diagnostic and therapeutic agents
  • Targets acidic diseased cells
Catalog No. 50-003-56204 Supplier Medchemexpress LLC Supplier No. HY144008B1MG
C8 PEG750 Ceramide is a lipid product designed to synthesize a lipid bilayer carrier. It enables the selective delivery of various diagnostic and therapeutic agents to acidic diseased cells.
  • Synthesizes a lipid bilayer carrier
  • Facilitates selective delivery of diagnostic and therapeutic agents
  • Targets acidic diseased cells