Learn More
CRYOCRATE C80EZ INSECT CRYOMEDIA 100ML
Catalog No. nc1621336
Encompass_Preferred
C80EZ® Insect is intended for the cryopreservation of insect cells across an array of types and applications (e.g., entomological research, protein expression). The media uniquely achieves predominant formation of nanoscale cubic ice during freezing, in contrast to the formation of large hexagonal ice in traditional cryopreservation media. The nanoscale ice attenuates cell mechanical damage due to extracellular ice formation and significantly improves the post-thaw viability and functionalities of cell types that traditionally survive poorly from cryopreservation. Use of C80EZ Insect eliminates the need for animal/human sera or antifreeze proteins.
Catalog No. NC1621336 Supplier CRYOCRATE Supplier No. 601001100
C80EZ® Insect is intended for the cryopreservation of insect cells across an array of types and applications (e.g., entomological research, protein expression). The media uniquely achieves predominant formation of nanoscale cubic ice during freezing, in contrast to the formation of large hexagonal ice in traditional cryopreservation media. The nanoscale ice attenuates cell mechanical damage due to extracellular ice formation and significantly improves the post-thaw viability and functionalities of cell types that traditionally survive poorly from cryopreservation. Use of C80EZ Insect eliminates the need for animal/human sera or antifreeze proteins.