Learn More
Aviva Systems Biology Corp C8B Antibody - middle region (ARP44210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C8B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human C8B. Alternate names: MGC163447ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53410