Learn More
ATCC C8-D30; Cerebellum; Mouse (Mus musculus)
Catalog No. 502381818
Encompass
C8-S [Astrocyte type II clone] is a cell line exhibiting neuronal-like morphology that was isolated from the cerebellum of an 8-day-old mouse. This cell line was deposited by B Pessac, and D Trisler. It can be used in immunology and neuroscience research.
Catalog No. 50-238-1818 Supplier ATCC Supplier No. CRL2534
C8-S [Astrocyte type II clone] is a cell line exhibiting neuronal-like morphology that was isolated from the cerebellum of an 8-day-old mouse. This cell line was deposited by B Pessac, and D Trisler. It can be used in immunology and neuroscience research.