Learn More
Aviva Systems Biology Corp TCIM Antibody - N-terminal region (ARP65059_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65059_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C8orf4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human C8orf4. Alternate names: MGC22806, TC-1, TC1, hTC-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-418