Learn More
Aviva Systems Biology Corp C8orf45 Antibody - middle region (ARP53264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C8orf45. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C8orf45. Alternate names: FLJ25692, C8orf45ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-819