Learn More
Aviva Systems Biology Corp C8orf70 Antibody - middle region (ARP32078_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32078_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C8orf70. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C8orf70. Alternate names: CGI-62, C8orf70, FAM164AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54447