Learn More
Aviva Systems Biology Corp C8orf77 Antibody - C-terminal region (ARP51723_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51723_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C8orf77. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C8orf77. Alternate names: MGC141859, C8orf77ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53665