Learn More
ATCC C8-S; Cerebellum; Mouse (Mus musculus)
Catalog No. 502381748
Encompass
C8-S [Astrocyte type II clone] is an astrocyte, type II cell line that was isolated from explant cultures of the cerebellum of an 8-day-old mouse. This cell line was deposited by B Pessac, and D Trisler. It can be used in immunology and neuroscience research.
Catalog No. 50-238-1748 Supplier ATCC Supplier No. CRL2535
C8-S [Astrocyte type II clone] is an astrocyte, type II cell line that was isolated from explant cultures of the cerebellum of an 8-day-old mouse. This cell line was deposited by B Pessac, and D Trisler. It can be used in immunology and neuroscience research.