Learn More
Aviva Systems Biology Corp C9orf127 Antibody - N-terminal region (ARP41900_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41900_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf127. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human C9orf127. Alternate names: NGX6, NAG-5, C9orf127, RP11-112J3.10ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58039