Learn More
Aviva Systems Biology Corp C9orf25 Antibody - middle region (ARP55478_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55478_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C9orf25. Alternate names: FLJ39031, bA573M23.5, C9orf25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61517