Learn More
Aviva Systems Biology Corp C9orf4 Antibody - N-terminal region (ARP47008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C9orf4. Alternate names: CG-6, CG6, C9orf4ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-59449