Learn More
Aviva Systems Biology Corp C9orf40 Antibody - middle region (ARP57082_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57082_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf40. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C9orf40. Alternate names: FLJ10110, FLJ25795ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-286-264