Learn More
Aviva Systems Biology Corp C9orf64 Antibody - N-terminal region (ARP60394_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60394_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf64. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C9orf64. Alternate names: MGC10999, RP11-575L7.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-031