Learn More
Aviva Systems Biology Corp C9orf68 Antibody - middle region (ARP56259_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56259_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf68. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C9orf68. Alternate names: FLJ10058, bA6J24.2, C9orf68ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61784