Learn More
Aviva Systems Biology Corp C9orf95 Antibody - N-terminal region (ARP57055_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57055_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf95. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C9orf95. Alternate names: FLJ20559, NRK1, RP11-235O14.2, bA235O14.2, C9orf95ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-260