Learn More
Aviva Systems Biology Corp STKLD1 Antibody - C-terminal region (ARP49089_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49089_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf96. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C9orf96. Alternate names: MGC43306, RP11-244N20.8, Sk521, SgK071ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-60018