Learn More
Medchemexpress LLC HY-103350 1mg Medchemexpress, CA-074 CAS:134448-10-5 Purity:>98%
SDP
Catalog No. 501968079
Encompass_Preferred
Medchemexpress, HY-103350 1mg CA-074 CAS:134448-10-5 CA-074 is a potent inhibitor of cathepsin B with a Ki of 2 to 5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8079 Supplier Medchemexpress LLC Supplier No. HY1033501MG
Medchemexpress, HY-103350 1mg CA-074 CAS:134448-10-5 CA-074 is a potent inhibitor of cathepsin B with a Ki of 2 to 5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.