Learn More
Apexbio Technology LLC CA-074 Me 147859-80-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062184
Encompass_Preferred
CA-074 Me (CAS 147859-80-1) is a cell-permeable methyl ester derivative of CA-074 functioning primarily as a selective inhibitor of cathepsin B It exhibits potent inhibitory activity against cathepsin B (IC50 36 3 nM) In cultured human gingival fibroblasts CA-074 Me effectively suppresses cathepsin B activity while moderately reducing combined cathepsin B and L activities Under strong reducing conditions (e g presence of DTT or GSH) CA-074 Me inhibits cathepsin L substantially Thus it is widely utilized in biomedical research to study lysosomal enzyme functions and roles of cathepsins in cellular processes
Catalog No. 50-000-62184 Supplier Apexbio Technology LLC Supplier No. A82395.1
CA-074 Me (CAS 147859-80-1) is a cell-permeable methyl ester derivative of CA-074 functioning primarily as a selective inhibitor of cathepsin B It exhibits potent inhibitory activity against cathepsin B (IC50 36 3 nM) In cultured human gingival fibroblasts CA-074 Me effectively suppresses cathepsin B activity while moderately reducing combined cathepsin B and L activities Under strong reducing conditions (e g presence of DTT or GSH) CA-074 Me inhibits cathepsin L substantially Thus it is widely utilized in biomedical research to study lysosomal enzyme functions and roles of cathepsins in cellular processes