Learn More
Selleck Chemical LLC CA-074 methyl ester
Supplier: Selleck Chemical LLC S742025MG
CA-074 methyl ester (CA-074 Me Cathepsin B Inhibitor IV) is a membrane-permeable derivative of CA-074 and acts as an irreversible cathepsin B inhibitor