Learn More
Medchemexpress LLC CA-074 methyl ester | 147859-80-1 | 397.47 | C19H31N3O6 | 5 MG
SDP
Catalog No. 5000213622
Encompass_Preferred
CA-074 methyl ester (CA-074Me) is a cell-permeable, selective inhibitor of Cathepsin B used in research on neuroprotection, cancer, and inflammatory processes. It is supplied as a powder with recommended storage conditions for both powder and solvent preparations, and it dissolves to form clear solutions in common organic solvents.
  • Selective Cathepsin B inhibition.
  • Cell-permeable methyl ester form.
  • Useful in neuroprotection, cancer, and inflammation studies.
  • Available in small research quantities including 5 mg.
  • Stable as powder at -20°C and in solvent under recommended conditions.
Catalog No. 50-002-13622 Supplier Medchemexpress LLC Supplier No. HY1003505MG
CA-074 methyl ester (CA-074Me) is a cell-permeable, selective inhibitor of Cathepsin B used in research on neuroprotection, cancer, and inflammatory processes. It is supplied as a powder with recommended storage conditions for both powder and solvent preparations, and it dissolves to form clear solutions in common organic solvents.
  • Selective Cathepsin B inhibition.
  • Cell-permeable methyl ester form.
  • Useful in neuroprotection, cancer, and inflammation studies.
  • Available in small research quantities including 5 mg.
  • Stable as powder at -20°C and in solvent under recommended conditions.