Learn More
Medchemexpress LLC CA-074 methyl ester | 147859-80-1 | MFCD00797532; MFCD03452890 | 397.47 | C19H31N3O6 | 25 MG
SDP
Catalog No. 5000213620
Encompass_Preferred
CA-074 methyl ester is a membrane-permeable, selective inhibitor of cathepsin B used in biochemical and cellular research to probe cathepsin B-mediated pathways. The methyl ester derivative improves cell permeability compared with the parent acid, enabling intracellular studies and functional assays.
  • Selective cathepsin B inhibition for targeted protease studies.
  • Membrane-permeable derivative suitable for cellular assays.
  • Useful in studies of neuroprotection, cancer, and inflammation.
  • Solid powder form with recommended storage at -20°C for long-term stability.
  • CAS: 147859-80-1; molecular weight 397.47 g/mol.
Catalog No. 50-002-13620 Supplier Medchemexpress LLC Supplier No. HY10035025MG
CA-074 methyl ester is a membrane-permeable, selective inhibitor of cathepsin B used in biochemical and cellular research to probe cathepsin B-mediated pathways. The methyl ester derivative improves cell permeability compared with the parent acid, enabling intracellular studies and functional assays.
  • Selective cathepsin B inhibition for targeted protease studies.
  • Membrane-permeable derivative suitable for cellular assays.
  • Useful in studies of neuroprotection, cancer, and inflammation.
  • Solid powder form with recommended storage at -20°C for long-term stability.
  • CAS: 147859-80-1; molecular weight 397.47 g/mol.