Learn More
Medchemexpress LLC CA-074 methyl ester | 147859-80-1 | MFCD00797532; MFCD03452890 | 397.47 g·mol⁻¹ | C19H31N3O6 | 1 ML
SDP
Catalog No. 5000214295
Encompass_Preferred
CA-074 methyl ester is a membrane-permeable, selective inhibitor of the cysteine protease cathepsin B, used in vitro and in vivo to probe cathepsin B-related biology. It exhibits reported neuroprotective, anti-cancer, and anti-inflammatory activities, and is suitable for preparing concentrated DMSO stocks or formulated in multiple in vivo solvent systems.
Catalog No. 50-002-14295 Supplier Medchemexpress LLC Supplier No. HY10035010MM 1ML
CA-074 methyl ester is a membrane-permeable, selective inhibitor of the cysteine protease cathepsin B, used in vitro and in vivo to probe cathepsin B-related biology. It exhibits reported neuroprotective, anti-cancer, and anti-inflammatory activities, and is suitable for preparing concentrated DMSO stocks or formulated in multiple in vivo solvent systems.