Learn More
Medchemexpress LLC CA-074 methyl ester | 147859-80-1 | MFCD03452890 | 397.47 g/mol | C19H31N3O6 | 50 MG
SDP
Catalog No. 5000213621
Encompass_Preferred
CA-074 methyl ester is a cell-permeable methyl ester prodrug of CA-074 that selectively inhibits Cathepsin B. It is used in biochemical and cellular research to investigate neuroprotective effects, cancer-related pathways, and inflammatory processes; intracellular esterases convert it to the active CA-074 inhibitor.
Catalog No. 50-002-13621 Supplier Medchemexpress LLC Supplier No. HY10035050MG
CA-074 methyl ester is a cell-permeable methyl ester prodrug of CA-074 that selectively inhibits Cathepsin B. It is used in biochemical and cellular research to investigate neuroprotective effects, cancer-related pathways, and inflammatory processes; intracellular esterases convert it to the active CA-074 inhibitor.