Learn More
Medchemexpress LLC CA-074 methyl ester | 147859-80-1 | MFCD03452890 | 99.2% | 397.47 g·mol⁻1 | C19H31N3O6 | 100 MG
SDP
Catalog No. 5000214847
Encompass_Preferred
CA-074 methyl ester is a selective, cell-permeable inhibitor of cathepsin B used in biochemical and cellular research to probe cathepsin-mediated pathways. It is commonly used in studies of neuroprotection, cancer biology, and inflammation where targeted inhibition of cathepsin B activity is required.
  • Selective inhibition of cathepsin B in biochemical assays.
  • Cell-permeable to enable intracellular targeting in cell-based studies.
  • High purity suitable for analytical and research applications.
  • Comes with supporting documentation including a datasheet and COA.
  • Available in multiple pack sizes to accommodate different experimental needs.
Catalog No. 50-002-14847 Supplier Medchemexpress LLC Supplier No. HY100350100MG
CA-074 methyl ester is a selective, cell-permeable inhibitor of cathepsin B used in biochemical and cellular research to probe cathepsin-mediated pathways. It is commonly used in studies of neuroprotection, cancer biology, and inflammation where targeted inhibition of cathepsin B activity is required.
  • Selective inhibition of cathepsin B in biochemical assays.
  • Cell-permeable to enable intracellular targeting in cell-based studies.
  • High purity suitable for analytical and research applications.
  • Comes with supporting documentation including a datasheet and COA.
  • Available in multiple pack sizes to accommodate different experimental needs.