Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CA-074 methyl ester | 147859-80-1 | MFCD03452890 | 99.3% | 397.47 | C19H31N3O6 | 10 MG
SDP

Catalog No. 5000214333
Encompass_Preferred

CA-074 methyl ester is a cell-permeable, selective inhibitor of cathepsin B used in biochemical and cellular research. The methyl ester derivative improves cell permeability compared with the parent acid, facilitating studies of cathepsin B-dependent pathways in neuroprotection, cancer, and inflammation models.

  • Selective cathepsin B inhibitor
  • Cell-permeable methyl ester derivative
  • High purity (99.34%)
  • Molecular weight 397.47 g/mol
  • Molecular formula C19H31N3O6
  • Available in multiple small-package sizes suitable for research use

Catalog No. 50-002-14333 Supplier Medchemexpress LLC Supplier No. HY10035010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CA-074 methyl ester is a cell-permeable, selective inhibitor of cathepsin B used in biochemical and cellular research. The methyl ester derivative improves cell permeability compared with the parent acid, facilitating studies of cathepsin B-dependent pathways in neuroprotection, cancer, and inflammation models.

  • Selective cathepsin B inhibitor
  • Cell-permeable methyl ester derivative
  • High purity (99.34%)
  • Molecular weight 397.47 g/mol
  • Molecular formula C19H31N3O6
  • Available in multiple small-package sizes suitable for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.