Learn More
Medchemexpress LLC HY-100350 1mg Medchemexpress, CA-074 methyl ester CAS:147859-80-1 Purity:>98%
SDP
Catalog No. 501931781
Encompass_Preferred
Medchemexpress, HY-100350 1mg CA-074 methyl ester CAS:147859-80-1 CA-074 methyl ester is a specific inhibitor of Cathepsin B , which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1781 Supplier Medchemexpress LLC Supplier No. HY1003501MG
Medchemexpress, HY-100350 1mg CA-074 methyl ester CAS:147859-80-1 CA-074 methyl ester is a specific inhibitor of Cathepsin B , which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.