Learn More
Cayman Chemical CA-074 methyl ester, Purity: ≥98%, CAS: 147859-80-1, 1 mg
SDP
Catalog No. 502966264
Encompass_Preferred
A cell-permeable inhibitor of cathepsin B and L in vivo and in whole cells; used to study the role of cathepsin B in neurodegeneration, cancer cell metastasis, necrosis, and rheumatoid arthritis
Catalog No. 50-296-6264 Supplier Cayman Chemical Supplier No. 184691 MG
A cell-permeable inhibitor of cathepsin B and L in vivo and in whole cells; used to study the role of cathepsin B in neurodegeneration, cancer cell metastasis, necrosis, and rheumatoid arthritis